Check Order Status
Vacuum and Pressure Bulbs
Actual product may vary
Fisherbrand
Vacuum and Pressure Bulbs
With vacuum and pressure PE valves. Deutsch Neumann™ Vacuum and Pressure Bulbs are manufactured from soft PVC.
Have Questions?
Catalog number
FKS10343791
Price (SGD)
56.00
EA
Add to cart
Price (SGD)
56.00
EA
Add to cart
Reddish-brown in color
With rigid connectors
Non-return valves ensure unidirectional flow
Specifications
Color
Brown
Material
PVC
Type
Vacuum Pressure Bulb
For Use With (Equipment)
For orsatapparat