Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Actual product may vary
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Fisherbrand
Nalgene™ BioTransport Carrier
Easily view contents with the Thermo Scientific™ Nalgene™ BioTransport Carrier.
Have Questions?
Catalog NumberMaterial
FKS10340301Polycarbonate
Catalog number FKS10340301
Price (SGD)
329.00
EA
Add to cart
Material:
Polycarbonate
Price (SGD)
329.00
EA
Add to cart
Transport racks containing tubes or sample holders filled with potentially infectious materials using the Thermo Scientific™ Nalgene™ BioTransport Carrier, which offers excellent visibility of contents.
  • Polycarbonate carrier is tough and break resistant
  • Easy to grasp, molded-in handles
  • Silicone gasket seals tightly
  • Clamps securely hold carrier closed and assure leak-resistant seal
  • Stainless-steel handle available separately (7136-0001)
  • For accidental spills inside of carrier, autoclave with clamps closed
To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Product TypeBioTransport Carrier
Certifications/ComplianceMeets OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
DescriptionBioTransport Carrier
AutoclavableAutoclavable
MaterialPolycarbonate
No. per Case4
No. per Pack1
Dimensions (L x W x H)14.5 x 7.3 x 6.687 in. (36.8 x 18.4 x 17.0cm)
Length (Metric)36.8 cm
Length (English)14.5 in.
Width (Metric)18.4 cm
Width (English)7.3 in.
Height (Metric)17.0 cm
Height (English)6.687 in.
Unit SizeEA