Isopore™ Membrane Filters, 0.4 μm
Isopore™ Membrane Filters, 0.4 μm
Actual product may vary
Fisherbrand
Isopore™ Membrane Filters, 0.4 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Color
FKS103399400.4 μm47 mmBlack
FKS100745300.4 μm25 mmBlack
Catalog number FKS10339940
Price (SGD)
3,637.00
PK
Add to cart
Pore Size:
0.4 μm
Diameter (Metric):
47 mm
Color:
Black
Price (SGD)
3,637.00
PK
Add to cart
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
AutoclavableNot Autoclavable
Bubble Point29 psi
ColorBlack
Filter TypeScreen Filter
MaterialPolycarbonate
Pore Size0.4 μm
SterilityNon-sterile
Flow RateAvg. 49 mL/min x cm2 (typical results @ 10 psi);18 mL/min x cm2/psi
For Use With (Application)Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Gas Flow Rate10.0 L/min/cm2
HydrophobicityHydrophilic
Quantity100/Pk.
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)47 mm
Unit SizePK