Check Order Status
96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color:
Clear;
Sterility:
Sterile;
Lid:
With Lid
Have Questions?
Change view
Catalog Number
Product Type
Volume (Metric) Working
FKS10334791
Microplate
1.5 to 2.2 mL
CRG353072
Microplate
1.5 to 2.2 mL
FIS08-772-2C
Microplate
1.5 to 2.2 mL
3 Options
Catalog number
FKS10334791
Price (SGD)
450.00
PK
Add to cart
Product Type:
Microplate
Volume (Metric) Working:
1.5 to 2.2 mL
Price (SGD)
450.00
PK
Add to cart
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
Opaque walls prevent well-to-well cross-talk.
Optically clear flat bottom permits direct microscopic viewing.
Crystal-grade virgin polystyrene
Sterilized by gamma irradiation
Nonpyrogenic
Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
Convenient, peel-open medical-style packaging for individual product presentation
Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Falcon 96, 384, and 1536 well microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates
Specifications
Format
Standard
Lid
With Lid
Culture Area
0.32 cm
2
Array
8 x 12
Packaging
1 Pk., 50 Cases
Binding Type
TC-Treated
Color
Clear
Product Type
Microplate
Material
Polystyrene
No. of Wells
96
Volume (Metric) Working
1.5 to 2.2 mL
Sterility
Sterile
Surface Treatment
Cell Culture
Total Volume (Metric) Well
0.37 mL
Well Shape
Flat Bottom (F-well)
No. per Case
50/Cs.
No. per Pack
1/Pk.
Quantity
50/Cs.