Nunc™ Microplate Lids
Nunc™ Microplate Lids
Actual product may vary
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Fisherbrand
Nunc™ Microplate Lids
Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityCondensation RingsCut-off Corners
FKS1033431196-well PlateSterileYesLong Side
263339
also known as 26333 9
96-well PlateNon-sterileYesLong Side
26412296-well PlateSterileYesLong Side
FIS12-565-7096-well PlateSterileYesLong Side
Catalog number FKS10334311
Price (SGD)
367.00
PK
Add to cart
Plate Compatibility:
96-well Plate
Sterility:
Sterile
Condensation Rings:
Yes
Cut-off Corners:
Long Side
Price (SGD)
367.00
PK
Add to cart
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications
ColorClear
Dimensions (L x W x H)5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
MaterialPolystyrene
SterilitySterile
No. of Wells96
Height (Metric)7.9 mm
Length (English)5 in.
Length (Metric)127 mm
Condensation RingsYes
Height (English)0.3 in.
No. per Case50
No. per Pack1
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Evaporation BarrierYes
Width (English)3.35 in.
Width (Metric)85 mm
Product TypePlate Lid
Product LineNunc
AdhesiveNone
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Plate Compatibility96-well Plate
Cut-off CornersLong Side
Unit SizePK