Check Order Status
Bottle Holder for Buerkle™ Telescopic Rods
Actual product may vary
Fisherbrand
Bottle Holder for Buerkle™ Telescopic Rods
Holds all bottles up to 88 mm (dia.). Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10333921
750 mL
Catalog number
FKS10333921
Price (SGD)
312.00
EA
Add to cart
Capacity (Metric):
750 mL
Price (SGD)
312.00
EA
Add to cart
With universal quick tightening strap
Polypropylene/PA
For industry and water treatment
Specifications
Product Type
Water Sampler
Material
Polypropylene, Polyamide
Includes
Universal quick tightening strap
Capacity (Metric)
750 mL