epT.I.P.S.™ Standard Pipette Tips
epT.I.P.S.™ Standard Pipette Tips
Actual product may vary
Fisher Catalogue
epT.I.P.S.™ Standard Pipette Tips
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)ColorPackaging
FKS103336821000 to 10,000 μL165 mmTurquoise200 Tips (2 Bags x 100 Tips)
FKS10157291100 to 5000 μL120 mmPurple500 Tips (5 Bags x 100 Tips)
FKS101991711000 to 10,000 μL243 mmTurquoise200 Tips (2 Bags x 100 Tips)
Catalog number FKS10333682
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1000 to 10,000 μL
Length (Metric):
165 mm
Color:
Turquoise
Packaging:
200 Tips (2 Bags x 100 Tips)
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
  • Minimal attachment and ejection forces with the highest level of tightness
  • The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • Fine graduations make visual checks of the pipetting volume even easier
  • Fine retracted or extended tips simplify work with small volumes
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
Specifications
Volume (Metric)1000 to 10,000 μL
FilteredNon-filtered
ColorTurquoise
AutoclavableAutoclavable
Length (English)6.5 in.
Length (Metric)165 mm
CompatibilityUniversal
FormatBulk
Tip StyleUniversal
Packaging200 Tips (2 Bags x 100 Tips)
Quantity200 Pk.
Product TypePipette Tips
For Use With (Equipment)Eppendorf Pipetter
GraduatedYes
Product LineepT.I.P.S.
GraduationsFine
Unit SizePK