CellBIND™ Surface Cell Culture Flasks
CellBIND™ Surface Cell Culture Flasks
Actual product may vary
Fisherbrand
CellBIND™ Surface Cell Culture Flasks
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Have Questions?
Catalog number FKS10327342
Price (SGD)
2,807.00
PK
Add to cart
Price (SGD)
2,807.00
PK
Add to cart
  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested
  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System
Specifications
BarcodeNo
Capacity (English)2.02 oz.
Closure ColorOrange
Closure MaterialHDPE
Capacity (Metric)60 mL
MaterialPolystyrene
For Use With (Application)For optimal cell attachment
IncludesLot numbers for ease in traceability
Neck StyleCanted
ShapeRectangular
Surface CoatingCorning Cellbind
GraduatedYes
SterilitySterile
Closure TypeVented
Unit SizePK