1x1 image pixel for data collection
Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Actual product may vary
Fisherbrand
Thermo/PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle
Features Diamond Syringe Technology which results in improved solvent resistance, wide temperature ranges, and increased operational smoothness. SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle are used for CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus, AS2000, AS3000, AS200/800 Autosamplers.
Have Questions?
Catalog NumberVolume (Metric)Needle GaugeDiameter (Metric) Outer NeedleModel
FKS1032701225 μL23 G0.63 mm25F-C/T-GT-5/0.63C
Catalog number FKS10327012
Price (SGD)
460.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Volume (Metric):
25 μL
Needle Gauge:
23 G
Diameter (Metric) Outer Needle:
0.63 mm
Model:
25F-C/T-GT-5/0.63C
Price (SGD)
460.00
EA
Add to cart
  • Improved solvent resistance
  • Wide temperature ranges up to 150°C
  • Increased operational smoothness
  • Designed specifically for the PAL
  • 22 gauge needles are 0.72mm OD
  • 23 gauge needles are 0.63mm OD
  • 26 gauge needles are 0.47mm OD
  • Accuracy and Reproducibility;  ±1% (dispensed volume) (±2% for 0.5μL syringes)
  • Cone shaped (also known as point style AS for autosampler), are designed to withstand multi injection demands and improves septum lifetime when used with an autosampler
  • Gas tight plunger, where applicable, is made of PTFE
Specifications
Needle Gauge23 G
Diameter (English) Inner Needle0.009 in.
Diameter (Metric) Inner Needle0.24 mm
Diameter (English) Outer Needle0.024 in.
Diameter (Metric) Outer6.5 mm
Diameter (Metric) Outer Needle0.63 mm
For Use With (Application)Gas chromatography
Length (English) Needle1.96 in.
Product TypeGas-tight Autosampler Syringe
Model25F-C/T-GT-5/0.63C
Quantity1/Pk.
Volume (Metric)25 μL
Needle Point StyleCone
For Use With (Equipment)CTC, LEAP, CombiPAL, GC PAL
Connection TypeFixed
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link