Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Actual product may vary
Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Absorb spills and cushion breakable items with long-lasting, waterproof, skid-resistant and chemical-resistant surface protectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensionsLxWDescription
FKS10325861Mat
FIS14-206-37250 ft. Dispenser Pk. Roll
74018-00Mat
FIS14-206-38Mat
FKS10182361Mat
FIS14-206-58Table Mat
74050-00Roll
74218-00Mat
FIS14-206-59Mat
Catalog number FKS10325861
Price (SGD)
682.00
PK
Add to cart
Description:
Mat
Price (SGD)
682.00
PK
Add to cart
Absorb spills and cushion breakable items with Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors. They are long-lasting, waterproof, skid-resistant and chemical-resistant. The cloth-like top layer will not fall apart even when wet.
  • Cushioned surface minimizes breakage of glass labware when dropped or tipped over
  • Sheets lie flat and are skid resistant
  • Low-pile flat surface provides stability for glassware that is likely to tip
  • Polyethylene backing resists harsh chemicals and protects benchtop from spills and corrosion
  • Super Versi-Dry quickly absorbs up to 1050mL/m2 to quickly absorb spills with thousands of Thermo Scientific Thirsty Cells
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Product TypeSurface Protector
Absorbancy1050 mL/m2
Length (English)18 in.
Length (Metric)46 cm
Width (English)20 in.
Width (Metric)51 cm
DescriptionMat
No. per Case100
No. per Pack100
Item DescriptionVersi-Dry Super Absorbant Mat 18 in. x 20 in.; 100 mats/case
Unit SizePK