SenTix™ 41 pH Electrode
SenTix™ 41 pH Electrode
Actual product may vary
Fisherbrand
SenTix™ 41 pH Electrode
Use these pH electrodes for portable measurement and routine measurement in the lab. WTW™ SenTix™ 41 pH Electrodes are ideal for applications where reliable measured values are needed under adverse conditions.
Have Questions?
Catalog NumberConnector Types
FKS10322911Fixed Cable
Catalog number FKS10322911
Price (SGD)
1,239.00
EA
Add to cart
Connector Types:
Fixed Cable
Price (SGD)
1,239.00
EA
Add to cart
pH electrodes with gel electrolyte are ideal for portable measurement but also for routine measurement in the lab. The electrodes do not require any maintenance besides occasional cleaning and are therefore especially economical. For demanding measurements in the lab, there are SenTix™ electrodes with 3 mol/l KCl liquid electrolyte, easy to clean glass shaft and platinum diagphragm. These electrodes can also be used in difficult samples. And if you need an electrode with liquid electrolyte for portable measuring: The SenTix™ 51/52 with plastic shaft, integrated temperature sensor and ceramic diaphragma masters nearly every measuring task, even in the field.
Specifications
Connector TypesFixed Cable
JunctionFiber
MaterialPlastic Shaft, Fiber Diaphragm
pH Range0 to 14
Temperature (Metric) Operating0°C to 80°C
TypeSenTix™ pH Electrode
For Use With (Equipment)Vario pH meter
IncludesDIN, +banana electrode plug, 1m cable
ModelSenTix™ 41
Probe30kilohms Built-in NTC
Storage RequirementsImmerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature (Metric) Range0°C to 80°C
Length (Metric) Cable1 m
Length (Metric) Stem120 mm
Unit SizeEA