Fluoropore™ Membrane Filters, 0.45 μm
Fluoropore™ Membrane Filters, 0.45 μm
Actual product may vary
Fisherbrand
Fluoropore™ Membrane Filters, 0.45 μm
Hydrophobic membrane with laminated backing for easier handling
Have Questions?
Catalog NumberThickness (Metric)
FKS10320071109 μm
Catalog number FKS10320071
Price (SGD)
2,399.00
PK
Add to cart
Thickness (Metric):
109 μm
Price (SGD)
2,399.00
PK
Add to cart
Hydrophobic membrane with laminated backing for easier handling
Our Fluoropore™ membrane is a hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve the handling characteristics of the filter for normal applications. Fluoropore™ membranes provide broad chemical compatibility, high flow rates and consistency. (Please note that catalogue numbers containing in.FHUP in. are unlaminated, or produced without a backing.)Applications: Fluoropore™ Membrane: Clarifying Acids, Bases and Solvents; Air Monitoring; Filtering and Venting Gases; UV Spectroscopy
Specifications
AutoclavableNot Autoclavable
Bubble Point7 psi
ColorWhite
Filter TypeScreen Filter
MaterialPTFE
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate60 mL/min x cm2
For Use With (Application)Clarification of Acids, Bases and Solvents; Clarification of Alcohol Solutions; UV Spectroscopy.
Gas Flow Rate8.0 L/min/cm2
HydrophobicityHydrophobic
Quantity100/Pk.
Thickness (Metric)109 μm
TypeFilter Membranes
WettabilityHydrophobic
Diameter (Metric)47 mm
Unit SizePK