LCD Display Thermo-Hygrometer
LCD Display Thermo-Hygrometer
Actual product may vary
Fisherbrand
LCD Display Thermo-Hygrometer
Use this thermohygrometer for a variety of applications. Testo™ 608-H1 Thermohygrometers provide a continuous display of current temperature and humidity values.
Have Questions?
Catalog number FKS10316802
Price (SGD)
473.00
EA
Add to cart
Price (SGD)
473.00
EA
Add to cart
  • Continuous measurement and display of temperature and relative humidity
  • Calculation and display of the dew point
  • Large, clear display
  • Capacitive humidity sensor with long-term stability for many years of measurement
Specifications
TypeThermohygrometer
Resolution0.001
Temperature Range (Metric)0°C to 50°C
Display TypeLCD
Weight (Metric)168 g
Voltage9 V
For Use With (Application)For measure and display the temperature and relative humidity continuously
Unit SizeEA