Round-Bottom Polypropylene Test Tubes With Cap
Round-Bottom Polypropylene Test Tubes With Cap
Actual product may vary
Fisherbrand
Round-Bottom Polypropylene Test Tubes With Cap
Manufactured from polypropylene for excellent chemical and thermal resistance. Falcon™Round-Bottom Polypropylene Tubes are tested to withstand 3000 x g centrifugation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Packaging
FKS103147915 mL75 mm25/Pk.; 500/Cs.
CRG3520635 mL75 mm25/Pk.; 500/Cs.
FIS14-959-11A5 mL75 mm25/Pk.; 500/Cs.
CRG35205914 mL100 mm25/Pk.; 500/Cs.
FIS14-959-11B14 mL100 mm25/Pk.; 500/Cs.
Catalog number FKS10314791
Price (SGD)
578.00
PK
Add to cart
Capacity (Metric):
5 mL
Length (Metric):
75 mm
Packaging:
25/Pk.; 500/Cs.
Price (SGD)
578.00
PK
Add to cart
  • Widely used and referenced in laboratory protocols
  • Dual position snap-cap prevents sample loss with ease of handling
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352002 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability
  • Dual-position snap caps, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 3,000 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • 17 x 100 mm tubes have blue printed graduations and white writing patch
  • Tube dimensions and volumes are approximate
Specifications
Closure ColorNatural
Closure MaterialPolyethylene
ColorClear
Description3000 x G
Capacity (Metric)5 mL
Length (Metric)75 mm
MaterialPolypropylene
DisposableSingle-use
IncludesSnap cap
Packaging25/Pk.; 500/Cs.
SterilitySterile
GraduatedNo
Diameter (Metric)12 mm
Closure TypeSnap Cap
Product TypeTest Tube
Unit SizePK