Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Actual product may vary
Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Fisher Catalogue
Nalgene™ Polycarbonate Centrifuge Bottles
Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNominal CapacityFor Use With (Equipment)
FKS103139711000 mLFiberlite™ rotors
3122-0250
also known as 3122-0250C
250 mLFiberlite™ rotors
3122-0500
also known as NB3122-0500
500 mLFiberlite™ rotors
3122-10001000 mLFiberlite™ rotors
FIS05-579-251000 mLFiberlite™ rotors
Catalog number FKS10313971
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Nominal Capacity:
1000 mL
For Use With (Equipment):
Fiberlite™ rotors
Process large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles, the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Molded of transparent polycarbonate, these clear bottles are autoclavable and have excellent mechanical strength properties.
Molded of transparent polycarbonate for easy visualization of samples, these bottles are ideal for low-speed centrifugation of biological and chemical samples.
  • For added leakproof† assurance at maximum speeds or when spinning hazardous materials, use with polypropylene sealing caps assemblies; available for 250mL (DS3131-0038) and 1000mL (DS3132-0063) sizes
  • Cat. No. 3122-1010 designed for use with IEC rotors
  • For proper performance, bottles must be filled to at least 80% of total capacity
Alert:
When spinning a 250mL bottle in a Thermo Scientific Sorvall GSA rotor, the use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)1000 mL
Capacity (English)33.81 oz.
ColorClear
DescriptionPolycarbonate Centrifuge Bottles
MaterialPolycarbonate
No. per Case16
No. per Pack4
Nominal Capacity1000 mL
AutoclavableAutoclavable
GraduatedNo
Max. Force7100 x g
ShapeRound
Height (Metric)177.0 mm
Product TypeCentrifuge Bottle
For Use With (Equipment)Fiberlite™ rotors
Closure MaterialPolypropylene
Diameter (Metric)98.2 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK