Amicon™ Ultra-15 Centrifugal Filter Units
Amicon™ Ultra-15 Centrifugal Filter Units
Actual product may vary
Fisherbrand
Amicon™ Ultra-15 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOQuantity
FKS1031188230 kDa24Pack
FISUFC910096100 kDa96Pack
Catalog number FKS10311882
View Price:Sign in to see your account pricing. Need an account? Register with us today.
MWCO:
30 kDa
Quantity:
24Pack
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
Specifications
DisposableSingle-use
Material (Membrane)High-recovery Ultracel-PL Regenerated Cellulose
MWCO30 kDa
SterilityNon-sterile
Filtration Area7.6 cm2
For Use With (Application)Protein purification, protein concentration and desalting
Housing MaterialRegenerated Cellulose (RC)
IncludesTwo microcentrifuge tubes
Quantity24Pack
TypeCentrifugal Filters
Volume (Metric)15 mL
Length (Metric)121 mm
Diameter (Metric)29.7 mm
Unit SizePK