Check Order Status
Natural Polypropylene Pipette Tips
Actual product may vary
Fisherbrand
Natural Polypropylene Pipette Tips
Use these superior Socorex™ Natural Polypropylene Pipette Tips to cover a range between 0.1 μL and 10 mL. They are versatile and broadly compatible with other brands of pipettes.
Have Questions?
Catalog Number
Volume (Metric)
FKS10307402
5 mL
Catalog number
FKS10307402
Price (SGD)
328.00
PK
Add to cart
Volume (Metric):
5 mL
Price (SGD)
328.00
PK
Add to cart
Polypropylene
Micropipette and macropipette tip options
Autoclavable
Specifications
Volume (Metric)
5 mL
Filtered
Non-filtered
Color
Natural
Autoclavable
Autoclavable
Length (Metric)
12.32cm
Format
Bulk
Tip Style
Macro
For Use With (Equipment)
Socorex™ Acura™ 5mL
Material
Polypropylene