Check Order Status
HyClone™ Water, Molecular Biology Grade, Cytiva
Element Search
Structure Search
Fisherbrand
HyClone™ Water, Molecular Biology Grade, Cytiva
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
Have Questions?
Catalog Number
Quantity
FKS10307052
1000 mL
Catalog number
FKS10307052
Price (SGD)
-
Quantity:
1000 mL
Request bulk or custom format
Specifications
pH
5.0 to 7.0
Recommended Storage
2° to 30°C
Description
Molecular Biology Grade Water
Endotoxin
0.05EU/mL
Filtered Through
Filter Sterilized - .1μm
View more
Format: Liquid