Wheaton™ Glass Staining Dishes for 50 Slides
Wheaton™ Glass Staining Dishes for 50 Slides
Actual product may vary
Fisherbrand
Wheaton™ Glass Staining Dishes for 50 Slides
Holds up to 50 slides horizontally
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
FKS10306472Dish, Cover, Rack
FIS08-813CDish, Cover
FIS22-029567Dish
Catalog number FKS10306472
Price (SGD)
5,602.00
PK
Add to cart
Includes:
Dish, Cover, Rack
Price (SGD)
5,602.00
PK
Add to cart
  • Dish and cover are made from soda-lime glass
  • Dimensions: 3.25 L × 7.875 W × 4.125 in. H (80 × 200 × 105 mm)
Rack
  • Nontarnishable and withstands most staining solutions
  • Holds up to 50 slides horizontally
  • Hinged handle allows easy insertion and removal of slides
Specifications
Closure IncludedYes
Bottom ShapeRectangular
MaterialSoda Lime Glass
TypeStaining Dish
For Use With (Equipment)Glass staining dish for 50 slides
IncludesDish, Cover, Rack
Slide Count50 slides horizontally
Dimensions (L x W)18.5 x 8.8 cm (7.2 x 3.4 in.)
Length (English)7.2 in.
Length (Metric)185 mm
Width (English)3.4 in.
Width (Metric)88 mm
Depth (English) Inside2.5 in.
Depth (Metric)68 mm
Depth (Metric) Inside6.5 cm
Unit SizePK