Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Actual product may vary
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Fisher Catalogue
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapeVolume (Metric) WellFit for Purpose
FKS10304513Round, Conical Bottom (V-well)200 μLPCR Ready
FISAB-0796Round330 μLPCR Ready
Catalog number FKS10304513
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Well Shape:
Round, Conical Bottom (V-well)
Volume (Metric) Well:
200 μL
Fit for Purpose:
PCR Ready
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Features
  • Alphanumeric grid-referencing and industry standard footprint
  • Polypropylene
  • Sealing options: Adhesive Seals, Heat Seals and Cap Strips
  • Cleanroom manufactured
Applications
  • Sample storage
  • Sample resuspension
  • Cell culture
  • Assay setup: diluting and aliquoting
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
FormatStandard
LidWithout Lid
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
PackagingBulk
Binding TypeLow binding PP
Product LineAbgene
ColorNatural
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Skirt StyleFull
MaterialPolypropylene
Volume (Metric) Well200 μL
No. of Wells96
Volume (Metric) Working200 μL
SterilityCleanroom
Surface TreatmentNon-treated
Well ShapeRound, Conical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF5000 x g
No. per Case100
No. per Pack1
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposePCR Ready
Plate SizeANSI/ SLAS Standard
Unit SizePK