Samco™ Graduated Transfer Pipettes
Actual product may vary
Fisherbrand
Samco™ Graduated Transfer Pipettes
Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityGraduationsPackaging Type
FKS102917901.5 mLSterile0.1, 0.2, 0.3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-243.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS153678133.9 mLNon-sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
2225.8 mLSterile, Sterile0.25, 0.50, 0.75, 1 mLBulk 500/Pk 10Pk/Cs 5000/Cs
FKS123844687.5 mLNon-sterile0.5, 1.0, 1.5, 2.0, 2.5, 3.0 mL
212-20S3.9 mLSterile0.25, 0.50, 0.75, 1 mL20/Bag, 500/Pk., 4000/Cs.
FKS153078235.8 mLSterile0.25, 0.50, 0.75, 1 mLPack of 500
2257.7 mLNon-sterile1, 2, 3 mLBulk 500/Pk 10Pk/Cs 5000/Cs
225-1S7.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS22-030-3207.7 mLSterile1, 2, 3 mLIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
FIS13-711-507.7 mLSterile1, 2, 3 mL20/Bag, 500/Pk., 4000/Cs.
FIS13-711-431.5 mLNon-sterile0.1 , 0.2, 0.3 mLBulk 500/Pk 8Pk/Cs 4000/Cs
FKS1233447813.7 mLNon-sterile1, 2, 3, 4, 5 mLBulk 250/Pk 10Pk/Cs 2500/Cs
Catalog number FKS10291790
Price (SGD)
230.00
Pack
Volume (Metric):
1.5 mL
Sterility:
Sterile
Graduations:
0.1, 0.2, 0.3 mL
Packaging Type:
Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs
Price (SGD)
230.00
Pack
Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)0.8 mL
DescriptionTransfer Pipette, Pediatric, Graduated to 0.1mL, 0.2mL, 0.3mL
Diameter (Metric) Outer3.0 mm
MaterialLDPE
SterilitySterile
Drops/mL22
GraduatedYes
Graduations0.1, 0.2, 0.3 mL
Packaging TypeIndividually Wrapped 500/Pk 8Pk/Cs 4000/Cs
Volume (Metric)1.5 mL
Length (Metric)11.6 cm
TypeGraduated Transfer Pipet