Use these extremely long, fine and flexible tips for filling of microcapillaries. Eppendorf ™ Microloader™ Pipette Tips are ideal for micro-injection and where additional reach is needed.
Have Questions?
Catalog Number
Volume (Metric)
FKS10289651
0.5 to 20 μL
Catalog number FKS10289651
Price (SGD)
872.00
Pack
Volume (Metric):
0.5 to 20 μL
Price (SGD)
872.00
Pack
For filling microinjection capillaries
0.5μL to 20μL
Racked
Specifications
Volume (Metric)0.5 to 20 μL
For Use With (Application)For filling microinjection capillaries
FormatRack
For Use With (Equipment)Eppendorf Microinjection Capillaries 0.5 μL to 20 μL