LDPE Printed Slope Shoulder Wash Bottles
LDPE Printed Slope Shoulder Wash Bottles
Actual product may vary
Fisherbrand
LDPE Printed Slope Shoulder Wash Bottles
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
Have Questions?
Catalog NumberCapacity (Metric)Chemical LabelClosure Color
FKS10287674250 mLAcetoneRed
Catalog number FKS10287674
Price (SGD)
103.00
PK
Add to cart
Capacity (Metric):
250 mL
Chemical Label:
Acetone
Closure Color:
Red
Price (SGD)
103.00
PK
Add to cart
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
  • Labeled wash bottles for easy identification of contents
  • Slim design enables comfortable handling and enhanced ease of use
  • Fitted with color-coded polypropylene closures
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
Diameter (Metric) Outer Neck54mm
For Use With (Application)For easy identification of contents
IncludesRed Polypropylene closure
MaterialLDPE
No. per Pack5/PK
Closure ColorRed
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)152 mm
Product TypeWash Bottle
Chemical LabelAcetone
Closure MaterialPolypropylene
Diameter (Metric)152 mm
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK