BlauBRAND™ Compact Burette
BlauBRAND™ Compact Burette
Actual product may vary
BlauBRAND™ Compact Burette
BlauBRAND™ Compact Burette
Fisherbrand
BlauBRAND™ Compact Burette
With PTFE stopcock, fast to dismantle and easy to clean
Have Questions?
Catalog NumberCapacity (Metric)
FKS1028716210 mL
Catalog number FKS10287162
Price (SGD)
336.00
EA
Add to cart
Capacity (Metric):
10 mL
Price (SGD)
336.00
EA
Add to cart
  • The new Generation of Burettes from BRAND™
  • With PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • Boro 3.3 Glass
  • Removable PTFE stopcock
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • All BlauBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
  • Specifications
    Capacity (Metric)10 mL
    MaterialBorosilicate Glass
    Graduations0.02 mL
    Stopcock MaterialPTFE
    Tolerance±0.02 mL
    Volume (Metric) Reservoir10 mL
    TypeCompact Buret
    ClassClass AS
    Certifications/ComplianceDIN EN ISO 385
    Unit SizeEA