Diaphragm Pump
Diaphragm Pump
Actual product may vary
Fisherbrand
Diaphragm Pump
Use these chemically resistant pumps for applications involving aggressive solvents and acidic vapors. Fisherbrand™ Diaphragm Pumps and connection heads are carbon fiber reinforced to provide electrical conductivity and prevent electrostatic charging.
Have Questions?
Catalog number FKS10287097
Price (SGD)
4,814.00
EA
Add to cart
Price (SGD)
4,814.00
EA
Add to cart
  • Chemically resistant, ideal for applications involving aggressive solvents and acidic vapors
  • Replacement for rotary vane pumps and water aspirators, especially in rough vacuum applications where corrosive vapors are encountered
  • Pump heads are made from PTFE with carbon fibre reinforcing to handle corrosive and aggressive vapors without damage
  • Pumping chambers are hermetically separated from drive space, ensuring extended life of mechanical parts
  • Pumps use PFTE diaphragms sandwiched to strong non-wetted backing and PEEK valves, which are chemical resistance and extremely hard wearing when compared to elastomer valves
  • Gas ballast valve helps to preserve pump performance when working with condensable vaporset
    • Hose nozzles for 8mm I. D. tubing
    • Vibration damping feet
Specifications
Max. Vacuum8 mbar
Electrical Requirements230 V 50/60 Hz motor with Schuko and UK power leads
Dimensions (L x W x H)19.5 x 23.5 x 14.5 cm (7.6 x 9.25 x 5.7 in.)
MaterialPTFE
Weight (English)14.3 lb.
Weight (Metric)6.5 kg
Flow Rate16 L/min.
Pump TypeDiaphragm Vacuum Pump
ApplicationsVacuum Oven
Maximum Pressure<8 mbar
Unit SizeEA