Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Actual product may vary
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Fisherbrand
Orion™ ROSS™ Sure-Flow™ Thin Stem Epoxy Bodied Combination pH Electrode
Attain stable, drift-free measurements of challenging samples in narrow containers using a sure-flow electrode with its unique free-flowing liquid-to-liquid junction.
Have Questions?
Catalog NumberConnector Types
FKS10283111BNC
Catalog number FKS10283111
Price (SGD)
2,181.00
EA
Add to cart
Connector Types:
BNC
Price (SGD)
2,181.00
EA
Add to cart
Thermo Scientific™ Orion™ ROSS Semi-Micro Sure-Flow™ electrodes are built with a unique free-flowing liquid-to-liquid junction that simplifies measurements of dirty or viscous samples by helping to prevent clogging, making them easy to clean. They are available in epoxy bodies for ruggedness and best suited for use in small samples of strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
DescriptionSure-Flow pH Electrode
Connector TypesBNC
JunctionSure-Flow
MaterialEpoxy Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypepH Electrode
Filling Solution Type3M KCl
IncludesDetachable bulb guard, 60mL bottle of ROSS electrode fill solution
Internal ReferenceROSS
Precision0.01
Temperature (English) Operating32°F to 212°F
Tip TypeSemi-micro
Diameter (Metric) Cap16 mm
Unit SizeEA