Easy Grip Disposable Polystyrene Sterile Bottles
Easy Grip Disposable Polystyrene Sterile Bottles
Actual product may vary
Fisherbrand
Easy Grip Disposable Polystyrene Sterile Bottles
Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS10278951500 mL
FIS09-761-140150 mL
CRG430281250 mL
FIS09-761-4250 mL
FIS09-761-10500 mL
FIS09-761-111 L
FKS101242911 L
Catalog number FKS10278951
Price (SGD)
665.00
PK
Add to cart
Volume (Metric):
500 mL
Price (SGD)
665.00
PK
Add to cart
  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Corning low profile style has easy grip sides that facilitate handling
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
Specifications
DisposableSingle-use
MaterialPolystyrene
SterilitySterile
For Use With (Application)Applications: Storage of media, buffers and other aqueous solutions
IncludesOrange HDPE plug seal
Mouth4.5 cm
No. per Case24/Cs.
No. per Pack2/Pk.
Quantity24 Case
TypeGraduated Easy Grip Storage Bottle
Volume (Metric)500 mL
Unit SizePK