Check Order Status
PCR-Cooler
Actual product may vary
Fisherbrand
PCR-Cooler
Innovative solution for protecting, transporting and storing sensitive samples as well as for safely stopping reactions.
Have Questions?
Catalog Number
Color
FKS10275091
Pink
Catalog number
FKS10275091
Price (SGD)
385.00
EA
Add to cart
Color:
Pink
Price (SGD)
385.00
EA
Add to cart
Maintains a sample temperature of 0°C for over an hour and then changes color when the temperature has exceeded 7°C
Superior to an ice bath, it is easy to handle, space saving and free of contamination, which makes it the perfect preparation for PCR reactions
Temperature of single tubes is indicated by the color of the individual bores
Samples are prevented from freezing by the specially developed cone contour
Cooling aggregate in the universal 96 well format for 0.2mL and 0.5mL PCR tubes, for tube strips, plates and individual tubes
Specifications
For Use With (Application)
PCR Reaction Setup
Type
PCR Cooler
Color
Pink
Capacity (Metric)
0.2 mL
Holds
96