Acid Rinse
Acid Rinse
Actual product may vary
Fisherbrand
Acid Rinse
Ideal for use as a pre-wash for heavy soiling or as an acid rinse to remove all detergent traces from apparatus and prevent the deposition of metal salts. Not suitable for domestic use
Have Questions?
Catalog NumberVolume (Metric)
FKS102734205000 mL
Catalog number FKS10273420
Price (SGD)
-
Volume (Metric):
5000 mL
Formulated from a blend of polycarboxylic acids, aliphatic alcohols and non-ionic surfactants in an aqueous base and is totally water rinsable. Because of its acidic nature, not suitable for non-ferrous metals, polycarbonate plastics, aluminum or zinc. This product is not suitable for domestic applications. Ideal for use as a pre-wash for heavy soiling or as an acid rinse to remove all detergent traces from apparatus and prevent the deposition of metal salts.
Specifications
FormLiquid
pH3.3
AppearanceColorless
Melting Point-63°C
Viscosity10.50cps at 20°C
Volume (Metric)5000 mL
Unit SizeEA