Pointed Bottom Hollow Stoppers, Hexagonal Plate
Pointed Bottom Hollow Stoppers, Hexagonal Plate
Actual product may vary
Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Have Questions?
Catalog NumberStandard Taper Joint
FKS10261731Thread Size: 14/23 Cone NS
Catalog number FKS10261731
Price (SGD)
65.00
PK
Add to cart
Standard Taper Joint:
Thread Size: 14/23 Cone NS
Price (SGD)
65.00
PK
Add to cart
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
DescriptionHollow
Quantity10/Pk.
Standard Taper JointThread Size: 14/23 Cone NS
MaterialBorosilicate Glass
Product TypeHollow Stopper
Unit SizePK