Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Actual product may vary
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Fisherbrand
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Catalog NumberConnector Types
FKS10255313Mini-DIN
Catalog number FKS10255313
Price (SGD)
1,675.00
EA
Add to cart
Connector Types:
Mini-DIN
Price (SGD)
1,675.00
EA
Add to cart
Thermo Scientific™ Refillable Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes have an built-in ATC. They are engineered with an epoxy body for ruggedness and best suited for general purpose samples.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required 
Electrode fill solution
900011     4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900011     4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Descriptionmini DIN connector; Glass fiber junction; Refillable
Connector TypesMini-DIN
JunctionGlass Fiber
MaterialEpoxy
pH Range0 to 14
Temperature (Metric) Operating0°C to 90°C
TypepH/ATC Probe, Mini-DIN connector
For Use With (Application)ideal for field or plant use.
For Use With (Equipment)Orion pH Meter (field or plant-use)
Internal ReferenceAg/AgCl
Temperature (English) Operating32°F to 194°F
Length (English)4.7 in.
Length (Metric)120 mm
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
Unit SizeEA