Pyrex™ Borosilicate Glass Heavy Wall Rimmed Test Tubes
Pyrex™ Borosilicate Glass Heavy Wall Rimmed Test Tubes
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Heavy Wall Rimmed Test Tubes
Manufactured with borosilicate glass for excellent chemical and thermal shock resistance. Pyrex™ Borosilicate Glass Heavy Wall Rimmed Test Tubes are ideal for chemical and biological work.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FKS1025365016 mm125 mm15 mL
Catalog number FKS10253650
Price (SGD)
608.00
PK
Add to cart
Diameter (Metric):
16 mm
Length (Metric):
125 mm
Capacity (Metric):
15 mL
Price (SGD)
608.00
PK
Add to cart
Manufactured with borosilicate glass for excellent chemical and thermal shock resistance. Pyrex™ Borosilicate Glass Heavy Wall Rimmed Test Tubes are ideal for chemical and biological work.
  • Rimmed heavy walls: provide superior mechanical strength
  • Robust construction: reduces breakage, lowers replacement costs and improves user safety
  • Conforms to: ISO 4142
Specifications
Certifications/ComplianceISO 4142
Capacity (Metric)15 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
Quantity100 Pack
RimmedYes
Diameter (Metric)16 mm
Unit SizePK