MF-Millipore™ Membrane Filter, 0.45 μm, Gridded
MF-Millipore™ Membrane Filter, 0.45 μm, Gridded
Actual product may vary
Fisherbrand
MF-Millipore™ Membrane Filter, 0.45 μm, Gridded
Mixed cellulose esters (MCE) membrane, hydrophilic
Have Questions?
Catalog NumberDiameter (Metric)Color
FKS1024809025 mmWhite
Catalog number FKS10248090
Price (SGD)
829.00
PK
Add to cart
Diameter (Metric):
25 mm
Color:
White
Price (SGD)
829.00
PK
Add to cart
Mixed cellulose esters (MCE) membrane, hydrophilic
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
Bubble Point≥1.8 bar, air with water
ColorWhite
Filter TypeMembrane Filter
MaterialMCE
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate60 mL/min x cm2
Gas Flow Rate4 L/min/ cm2
HydrophobicityHydrophilic
Membrane TypeGridded
No. per Case100Pack
Quantity100/Pk.
Thickness (Metric)150 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)25 mm
Unit SizePK