HM-200 Series Multiposition Extraction Heater
HM-200 Series Multiposition Extraction Heater
Actual product may vary
Fisherbrand
HM-200 Series Multiposition Extraction Heater
The HM-200 Series Multiposition Extraction Heater allows liquids with different boiling points to be heated next to each other.
Have Questions?
Catalog NumberCapacity (Metric)For Use With
FKS10247524250 mLFlasks (6 x 250 mL)
Catalog number FKS10247524
Price (SGD)
9,386.00
EA
Add to cart
Capacity (Metric):
250 mL
For Use With:
Flasks (6 x 250 mL)
Price (SGD)
9,386.00
EA
Add to cart
  • Unique vented airflow ensures the case remains ''cool-to-the-touch.''
  • Heat your solutions simultaneously in each position.
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests.
  • Regulating the heat on these multiposition extraction mantles is simple using the built-in solid-state electronic controls.
  • The units accept round-bottom flasks ranging from 100 to 1000 mL.
  • Select an extraction heater depending on the size of your flask.
  • Extraction heaters feature an independent heater per mantle, allowing liquids with different boiling points to be heated alongside each other.
  • Each heater has its own control dial and on/off indicator light, and can reach up to a maximum element temperature of 450°C (842°F).
  • The control panel is safely separated from the heating elements by a well-ventilated air space.
  • A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater.
  • Powder-coated aluminum surfaces and polypropylene top cover ensure good chemical resistance and easy cleanup.
Specifications
Certifications/ComplianceCE
Capacity250mL (8.4 oz.)
DesignPolypropylene housing
Capacity (Metric)250 mL
Depth (English)10.25 in.
Depth (Metric)26 cm
For Use WithFlasks (6 x 250 mL)
Height (English)3.5 in.
Height (Metric)8.9 cm
Voltage230 V
Electrical Requirements230 V 50/60 Hz, 900 W (UK and EU plug)
ModelEME60250/CEB
Wattage900 W
Width (English)47.2 in.
Width (Metric)121 cm
Product TypeHeating Mantle
Temperature (Metric)Ambient to 450°C
Amperage3.9 A
Capacity (English)8.4 oz.
For Use With (Application)Heating solutions
Unit SizeEA