WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
Actual product may vary
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
Fisherbrand
WypAll™ L30 General Clean™ Heavy Duty Wipes BL 7426 , 1 Roll x 670
Extra wide jumbo roll industrial wiping paper is ideal for cleaning, soaking up industrial spills, and wiping oily or dirty parts and surfaces. The 3 ply blue paper towels feature Airflex™ technology, to deliver exceptional strength and absorbency. 1 cleaning roll x 670 blue paper sheets (670 sheets total).
Have Questions?
Catalog NumberDimensions (L x W)
FKS1024336238 x 37 cm
Catalog number FKS10243362
Price (SGD)
-
Dimensions (L x W):
38 x 37 cm
  • Each individual roll of L30 heavy duty wipes contains 670 blue, 3 ply wiping sheets
  • Industrial wipes designed for demanding, heavy-duty wiping tasks like soaking up spills and wiping oily parts and surfaces
  • Unique Airflex™ technology in every blue towel roll delivers exceptional strength and absorbency
  • Single use wiping paper in a jumbo blue roll format
  • Compatible with a variety of Kimberly-Clark Professional™ dispensers
Specifications
Product TypePerforated Dry Wiper Roll
Length (English)14.56 in.
Length (Metric)37.5 cm
Width (English)14.56 in
Width (Metric)37.5 cm
MaterialPaper
ColorBlue
Dimensions (L x W)38 x 37 cm
For Use With (Application)For routine cleaning work in the industry, for the cleaning of conveyor belts, to wipe large amounts of spilled oil or grease, for cleaning of the face and hands in patients, counteracts the spread of pathogens Use with oil or water
Packaging Quantity670 Sheets
Ply3-ply