1x1 image pixel for data collection
gasprofi 1 SCS micro Gas Burner
gasprofi 1 SCS micro Gas Burner
Actual product may vary
Fisherbrand
gasprofi 1 SCS micro Gas Burner
The smallest professional laboratory gas burner. Wld Tec™ Gasprofi 1 SCS Micro Gas Burner offers optimal safety and easy operation.
Have Questions?
Catalog number FKS10241202
Price (SGD)
3,076.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Price (SGD)
3,076.00
PK
Add to cart
  • Button or foot pedal operation
  • Gas inlet filter
  • Safety control system (SCS)
  • Removable and decomposable burner head
  • Housing, burner head, gas connection and controls made of stainless steel
  • DVGW approved
Specifications
Product TypeBunsen Burner
Gas TypeNatural Gas, Butane, Propane
Length (Metric)85 mm
MaterialStainless Steel
Height (Metric)49 mm
IncludesFoot Pedal and Button Function
Max. Temperature (Metric)1300°C (Natural Gas), 1350°C (Liquid Gas)
No. per Pack1 Ea.
Weight (Metric)700 g
Width (Metric)86 mm
Unit SizePK
Documents & Downloads
Certificates
Share catalog number, name or link