Quantofix™ Formaldehyde Test Strip Kits
Quantofix™ Formaldehyde Test Strip Kits
Actual product may vary
Fisherbrand
Quantofix™ Formaldehyde Test Strip Kits
Allows for the quick and easy determination of formaldehyde in solutions. Macherey Nagel™ Quantofix™ Formaldehyde Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Have Questions?
Catalog NumberColor Indicator
FKS10238390Beige to Blue/Violet
Catalog number FKS10238390
Price (SGD)
283.00
PK
Add to cart
Color Indicator:
Beige to Blue/Violet
Price (SGD)
283.00
PK
Add to cart
Allows for the quick and easy determination of formaldehyde in solutions. Macherey Nagel™ Quantofix™ Formaldehyde Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable result within one minute
  • Easily monitor closed cooling or heating circuits
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C
Specifications
Color IndicatorBeige to Blue/Violet
FormatStrip
For Use With (Application)Determination of Formaldehyde
Graduations0, 10, 20, 40, 60, 100, 200mg/L
Quantity100 Pack
Shelf Life2.5 Years
Storage Requirements30°C
Test Time1 min.
TypeSemi-qualitative Test Strip
Unit SizePK