PIPETMAN™ Classic Pipet
PIPETMAN™ Classic Pipet
Actual product may vary
Fisherbrand
PIPETMAN™ Classic Pipet
The PIPETMAN Classic line, the world standard in pipetting, offers a wide range of continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density.
Have Questions?
Catalog NumberVolume (Metric)
FKS102360721 to 10 μL
Catalog number FKS10236072
Price (SGD)
1,248.00
EA
Add to cart
Volume (Metric):
1 to 10 μL
Price (SGD)
1,248.00
EA
Add to cart
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability
Specifications
Accuracy±0.025, ±2.5μL
Compatible TipsD10, DL10, DF10ST, DFL10ST
Adjustable Tip SpacingNo
ColorRed
Dispensing SpeedManual
DisposableReusable
Grip TypeErgonomic
MaterialPolypropylene
ModelP10
Number of Channels1
Product LinePipetman Classic
ProgrammabilityNo
SterilityNon-sterile
TypePipette
Volume (Metric)1 to 10 μL
For Use With (Application)PCR, DNA Sequencing, Small Volume Dispensing of Aqueous Fluids of Moderate Viscosity and Density (Molecular Biology, Immunology)
Length (English)14.37 in.
Length (Metric)36.5 cm
Width (English)12.01 in.
Width (Metric)30.5 cm
Height (English)4.72 in.
Height (Metric)12 cm
Color CodeRed
Fit for PurposeCertified Quality
Unit SizeEA