Research™ plus EU-IVD Single Channel Variable Volume Pipette
Research™ plus EU-IVD Single Channel Variable Volume Pipette
Actual product may vary
Fisherbrand
Research™ plus EU-IVD Single Channel Variable Volume Pipette
The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
Have Questions?
Catalog NumberVolume (Metric)Color
FKS10234204100 to 1000 μLBlue
Catalog number FKS10234204
Price (SGD)
1,536.00
EA
Add to cart
Volume (Metric):
100 to 1000 μL
Color:
Blue
Price (SGD)
1,536.00
EA
Add to cart
The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (not available for 5 mL and 10 mL pipettes) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs to ensure contamination-free operation
Specifications
Certifications/ComplianceEU-IVD
Compatible TipsepT.I.P.S.™ 1 000 μL
AutoclavableAutoclavable
ColorBlue
DisposableReusable
For Use WithepT.I.P.S.™ 1000 μL
Number of Channels1
Product LineResearch™ Plus™
TypeSingle Channel Manual Pipette
Volume (Metric)100 to 1000 μL
For Use With (Application)Forward pipetting, Reverse pipetting, Removal of supernatants, Sample mixing, Phase extraction, Volume determination, Filling of plates, gels, and reaction vessels
Unit SizeEA