Short-Term Detector Tubes: Toluene
Short-Term Detector Tubes: Toluene
Actual product may vary
Fisherbrand
Short-Term Detector Tubes: Toluene
Detector tubes
Have Questions?
Catalog NumberComponents MeasuredTube TypeMeasurement RangeHumidity Range
FKS10222681TolueneToluene 5/b5–300ppmNot Applicable
Catalog number FKS10222681
Price (SGD)
671.00
PK
Add to cart
Components Measured:
Toluene
Tube Type:
Toluene 5/b
Measurement Range:
5–300ppm
Humidity Range:
Not Applicable
Price (SGD)
671.00
PK
Add to cart
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Specifications
Components MeasuredToluene
Humidity RangeNot Applicable
Measurement Range5–300ppm
No. of Strokes2/10
Quantity10
Response Time1 to 5 min.
Temperature Range (English)41°F to 77°F
Temperature Range (Metric)2° to 40°C
Tube TypeToluene 5/b
Unit SizePK