BioCoat™ #1 German Glass Coverslips, Round
BioCoat™ #1 German Glass Coverslips, Round
Actual product may vary
Fisherbrand
BioCoat™ #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Have Questions?
Catalog NumberDiameter (Metric)Surface Treatment
FKS1022077212 mmPoly-L Lysine
Catalog number FKS10220772
Price (SGD)
2,079.00
PK
Add to cart
Diameter (Metric):
12 mm
Surface Treatment:
Poly-L Lysine
Price (SGD)
2,079.00
PK
Add to cart
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Specifications
TypeSlide Coverslip
Surface TreatmentPoly-L Lysine
MaterialNo. 1 German Glass
Diameter (English)0.472 in.
For Use With (Application)Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy
Diameter (Metric)12 mm
No. per Case80
No. per Pack80
Unit SizePK