NAP™ Columns, NAP-5
NAP™ Columns, NAP-5
Actual product may vary
Fisherbrand
NAP™ Columns, NAP-5
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
Have Questions?
Catalog NumberQuantity
FKS1021828450 Pack
Catalog number FKS10218284
Price (SGD)
-
Quantity:
50 Pack
  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange
Specifications
For Use With (Application)Rapid and efficient purification of DNA and Oligonucleotides
IncludesComplete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product TypeNAP-5
Quantity50 Pack
Unit SizePK