Mini Magnifier
Mini Magnifier
Actual product may vary
Fisherbrand
Mini Magnifier
Provides hands-free 1.75X magnification of slides, plates or gels placed on the Mini Light Box II
Have Questions?
Catalog NumberMagnification
FKS102162041.75x
Catalog number FKS10216204
Price (SGD)
1,621.00
EA
Add to cart
Magnification:
1.75x
Price (SGD)
1,621.00
EA
Add to cart
  • Tilts for optimal viewing
  • Measures 10.5 x 14.3 x 17.2cm H
  • Also useful for viewing of x-ray films, stained gels, slides or text
Specifications
For Use With (Application)Magnified observation of petri dishes, slides, gels or text
For Use With (Equipment)Scienceware™ Mini Light Box II
Height (English)6.7 in.
Height (Metric)17.2 cm
Length (English)6.25 in.
Length (Metric)15.8 cm
Weight (English)13 oz.
Weight (Metric)368.5 g
Width (English)5.9 in.
Width (Metric)14.9 cm
Diameter (Englis) Lens5.5 in.
Diameter (Metric) Lens140 mm
Magnification1.75x
MaterialGlass Lens
Product TypeMagnifier, Mini
Unit SizeEA