PYREX™ Disposable Round-Bottom Rimless Glass Tubes
PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Actual product may vary
Fisherbrand
PYREX™ Disposable Round-Bottom Rimless Glass Tubes
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)
FKS1021256319 mL125 mm
FIS14-962-15F19 mL125 mm
FIS14-962-15A4 mL75 mm
FIS14-962-1C6 mL75 mm
FKS1001852223 mL150 mm
FIS14-962-15E15 mL100 mm
Catalog number FKS10212563
Price (SGD)
575.00
PK
Add to cart
Capacity (Metric):
19 mL
Length (Metric):
125 mm
Price (SGD)
575.00
PK
Add to cart
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ Disposable Round-Bottom Rimless Glass Tubes are designed for both tissue culture and general bacteriological work.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed. Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
Specifications
Capacity (Metric)19 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithFor tissue culture work and general bacteriological use
Label TypeNone
PackagingBulk
RimmedNo
SterilityNon-sterile
GraduatedNo
Diameter (Metric)16 mm
Product TypeCulture Tube
Unit SizePK