Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Actual product may vary
Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Fisherbrand
Nalgene™ Class B Polypropylene Copolymer Volumetric Flasks with Closure
Lighter in weight than glass, these flasks are break-resistant, offer excellent chemical resistance and protect against leakage and contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeLimit of Error
FKS1021256150 mL13 mm±0.10 mL
FIS10-198-50A50 mL13 mm±0.10 mL
FIS10-198-50B100 mL20 mm±0.16 mL
FKS10018780100 mL20 mm±0.16 mL
FIS10-198-50C200 mL20 mm±0.20 mL
FIS10-198-50D250 mL20 mm±0.24 mL
FIS10-198-50F1000 mL28 mm±0.60 mL
Catalog number FKS10212561
Price (SGD)
142.00
EA
Add to cart
Capacity (Metric):
50 mL
Closure Size:
13 mm
Limit of Error:
±0.10 mL
Price (SGD)
142.00
EA
Add to cart
Lighter in weight than glass, Thermo Scientific™ Nalgene™ ASTM Class B Polypropylene Copolymer Volumetric Flasks with Closure offer excellent chemical resistance and are calibrated To Contain and To Deliver at 20°C (68°F). These shatterproof flasks can play an important part in lab safety programs. Leakproof† protection when used with provided Nalgene closure.
Features:
  • Break resistant, excellent chemical resistance for years of dependable use in your lab
  • Individually hand calibrated, permanently engraved with a colored graduation line for ease in reading fluid levels
  • Oval marking spot
  • No meniscus to confuse readings
  • 200mL size has additional line at 203mL level for BOD and DO determinations
  • Meets ASTM E 288 and ISO 384 Class B tolerances for consistent quality and dependability
  • Autoclavable/Graduated/Leakproof† when used with Nalgene closure
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Warning: Autoclavable but accuracy will be affected. Before autoclaving, remove closure and set it on top without engaging threads.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (English)1.69 oz.
Certifications/ComplianceMeets ASTM E 288 and ISO 384 Class B tolerances for consistent quality and dependability
DescriptionFlask Volumetric PP 50mL
Capacity (Metric)50 mL
MaterialPPCO
Height (English)5.8 in.
Height (Metric)148 mm
Limit of Error±0.10 mL
No. per Case12
No. per Pack1
AutoclavableAutoclavable but will affect accuracy
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
ClassB
Bottom ShapeRound
Closure TypeScrew
Diameter (Metric) Outer46 mm
Weight (Metric)14 g
Culture AreaNot applicable
Volume (Metric) Working50 mL
Closure Size13 mm
Unit SizeEA