Borosilicate Glass Automatic Titrating Burette with Storage Bottle
Borosilicate Glass Automatic Titrating Burette with Storage Bottle
Actual product may vary
Fisherbrand
Borosilicate Glass Automatic Titrating Burette with Storage Bottle
For use in laboratory, electroplating industry, and water treatment applications. Burettes feature precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)
FKS1020823010 mL56 cm
Catalog number FKS10208230
Price (SGD)
302.00
EA
Add to cart
Capacity (Metric):
10 mL
Height (Metric):
56 cm
Price (SGD)
302.00
EA
Add to cart
  • Graduation according to Category B (Tolerances according to DIN EN ISO 385)
  • Adjusted for Ex
  • Burette made of borosilicate glass 3.3
  • Resistant to chemicals
  • Blue ring graduations
  • Left or right hand operation possible
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with the medium: PE, PP, natural rubber, borosilicate glass
Specifications
Accuracy0.045 mL
Capacity (Metric)10 mL
Length (Metric)56 cm
MaterialBorosilicate Glass
Graduations0.05 mL
For Use With (Application)Water Treatment Plants, Electroplating Industry, Field Analyses
Height (Metric)56 cm
IncludesMicro-Screw, Precise Discharge Jet, Sturdy Stand
ScaleBlue
Stopcock MaterialBorosilicate Glass, Polyethylene, Polypropylene, Rubber
TypeAutomatic Titrating Buret
Certifications/ComplianceDIN ISO 385
Unit SizeEA