Check Order Status
Glass Laboratory Sprayer
Actual product may vary
Fisherbrand
Glass Laboratory Sprayer
Used as an accessory in thin layer chromatography applications. Macherey-Nagel™ Glass Laboratory Sprayer includes a 100mL, clear glass conical flask with a sprayer-stopper assembly and rubber bulb/tubing.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10206621
100 mL
Catalog number
FKS10206621
Price (SGD)
661.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
661.00
EA
Add to cart
Specifications
Capacity (Metric)
100 mL
Color
Clear-Red
Material
Glass, Rubber
Quantity
1 Ea.
Type
Laboratory Sprayer
For Use With (Application)
TLC
Includes
Rubber bulb/tubing