HDPE Wide Neck 5L Bottle
HDPE Wide Neck 5L Bottle
Actual product may vary
Fisherbrand
HDPE Wide Neck 5L Bottle
Tested and certified to meet UN standards for the safe transportation of hazardous materials
Have Questions?
Catalog NumberCapacity (Metric)
FKS102031815 L
Catalog number FKS10203181
Price (SGD)
42.00
EA
Add to cart
Capacity (Metric):
5 L
Price (SGD)
42.00
EA
Add to cart
  • Supplied with tamper evident polypropylene cap
  • Moulded in carrying/pouring handle
  • Unapproved
  • Color: Clear
Specifications
Capacity (Metric)5 L
ColorClear
Diameter (Metric) Outer Neck45 mm
MaterialHDPE
Quantity1
Width (Metric)170mm
Certifications/ComplianceTested and certified to meet UN standards for the safe transportation of hazardous materials
Height (Metric)330 mm
Closure MaterialPolypropylene
Closure TypeTamper-evident
Unit SizeEA