Polypropylene Straight Connectors with Conical Nozzles
Polypropylene Straight Connectors with Conical Nozzles
Actual product may vary
Fisherbrand
Polypropylene Straight Connectors with Conical Nozzles
Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Straight Connectors with Conical Nozzles resist temperatures up to 135°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
FKS101982517 to 10 mm Hose
FKS1013816111 to 14 mm Hose
Catalog number FKS10198251
Price (SGD)
54.00
PK
Add to cart
For Use With:
7 to 10 mm Hose
Price (SGD)
54.00
PK
Add to cart
Cylindrical straight hose connector with conical nozzles
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
Specifications
Certifications/ComplianceEU Foodstuffs, RoHS
AutoclavableAutoclavable
MaterialPolypropylene
For Use With7 to 10 mm Hose
Packaging Quantity10/Pk.; 10/Scale
ShapeStraight
ColorWhite
Product TypeHose Tubing Connector
Unit SizePK