1x1 image pixel for data collection
Standard Duty WOB-L™ Piston Vacuum Pump: Model 2522
Standard Duty WOB-L™ Piston Vacuum Pump: Model 2522
Actual product may vary
Fisherbrand
Standard Duty WOB-L™ Piston Vacuum Pump: Model 2522
For non-chemical applications
Have Questions?
Catalog number FKS10191942
Price (SGD)
3,339.00
EA
Add to cart
Price (SGD)
3,339.00
EA
Add to cart
Wobbling piston wobbles providing air resistance on upward stroke expanding Teflon seal on piston and increases its efficiency for wobble action.
  • Oil free dry pump powered by WOB-L piston
  • Intake/Exhaust thread NPT: 1/4
Model 2522
  • Free air displacement: CFM (L/min) at 60Hz: 0.76 (22)
  • Ultimate pressure: 100 torr (133mbar)
  • Maximum pressure: 100psig
  • Maximum vacuum in Hg: 26
  • Motor: 0.12 hp (93w)
  • For pumping vapors of aqueous solutions including buffers but not for acidic, basic or organic vapors or gases
  • 50Hz units supplied with CE marking
Vacuum filtration, Vacuum oven, Vacuum chamber/Roughing; Dessication; Aspiration; Automation; Cell culture; Glove box
Specifications
Horsepower0.12 hp
Shipping Weight (English)15 lb.
Shipping Weight (Metric)6.6 kg
IncludesConvenient gauge read cut, pressure adjustment valve, protective separator jar, Euro (Schuko) plug
Certifications/ComplianceCE
Electrical Requirements230 V 50 Hz
Model2522
Weight (English)11.7 lb.
Weight (Metric)5.3 kg
For Use WithVacuum filtration, vacuum oven, vacuum chamber/roughing, desiccation, aspiration, automation, cell culture
Frequency50 Hz
Overall Height (English)10 in.
Overall Height (Metric)25.4 cm
Overall Length (English)8.1 in.
Overall Length (Metric)20.6 cm
Overall Width (English)8.8 in.
Pump TypeVacuum Pump
Maximum Pressure133 mbar
Voltage230 V
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link